BUCHI™ Receiving Flasks for Rotary Evaporators
Use these receiving flasks with any rotary evaporator. Buchi™ Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.
Manufacturer: BUCHI™ 000422
 View more versions of this product
Catalog No. 05-402-652
  Check Availability
Add to Cart
    Specifications
    Capacity (Metric) 100mL
    For Use With (Equipment) Rotary evaporator
    Product Type Receiving Flask
    Material Standard Glass
    Material (Coating) Uncoated
    Includes Ball joint KS-35
    Shape Round
    Volume (English) 100mL
  • Description & Specifications
Related Products
Product Content Feedback