Learn More
BUCHI™ Rotavapor™ R-210 Rotary Evaporator System w/Glassware A or C, Vacuum Pump, Vacuum Controller and Recirculating Chiller
Reliable and convenient operation
Manufacturer:  BUCHI™ 23021A01111
 View more versions of this product
Catalog No. 05-403-350
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
CE, CSA, UL
4000mL (135.2 oz.)
42cm
33.07 in.
16 to 18kg
100-120V
59cm
135.2 oz.
R-210
20° to 180°C
P+G Without coating
68° to 356°F
50 to 4000mL
100-120V 50/60Hz
V-850
3 Years
16.5 in.
840mm
50/60Hz
35.27 to 39.68 lb.
23.22 in.
29/32
4000mL
20 to 280rpm
Standard applications where height is limited
Heating bath, Recirculating Chiller, Evaporating flask (1 liter), Vacuum controller, Vacuum pump
Woulff bottle for vacuum controller
60w
A