Learn More
BUCHI™ Rotavapor™ R220 SE Industrial Evaporator Available on GSA/VA Contract for Federal Government customers only.
Easy handling ensures safer operation
Manufacturer:  BUCHI™ 118114C272
 View more versions of this product
Catalog No. 05-412-925
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications Description & Specifications
Specifications
CE, CSA, UL
20L
180°C
Water or oil up to 180°C
DB2 condensor, 20L evaporating flask, Stainless Steel heating bath
Spring loaded shut-off valve
230V
5 to 150rpm
Laboratory evaporation, pharmaceutical, chemical, environmental, evaporation feed, food, beverage, academia, education analysis
R220 Se Industrial Evaporator
356°F
20000mL