Learn More
BUCHI™ Rotavapor™ R220 SE Industrial Evaporator Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  BUCHI™ 118114C271
 View more versions of this product
Catalog No. 05-412-926
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications Description & Specifications
Specifications
UL/CSA, CE
70.86 in.
143.3 lb.
47.24 in.
13.5L/hr.
LCD
Electrical vacuum valve and cooling water valve
V-850
1800mm
65kg
220 to 240V
475.13 oz./hr.
5 to 150rpm
Safe and efficient fast distillation in pharmaceutical and chemical industry for easy distillable or drying products, heat sensitive products, reflux distillations, solvent recycling and foaming applications
2 Wire PT-1000
4200w