BUCHI™ Rotavapor™ RII Evaporation Systems

For quick, gentle and economical evaporation procedures without cutting corners

Manufacturer: BUCHI™ 2422V0

 View more versions of this product

Catalog No. 05-403-617

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided. After extensive use, coating on glassware may wear and loosen from surface, increasing risk of injury.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.


Required Accessories : Vacuum pump or system and associated connections.

    Specifications

    Certifications CE, CSA, UL
    Weight (English) 35.2 to 39.6 lb.
    Width (English) 23.62 in.
    Capacity (English) 135.25 oz.
    Capacity 4000mL (135.25 oz.)
    Height (Metric) 84cm
    Height (English) 33.07 in.
    Weight (Metric) 16 to 18kg
    Taper 24/40
    For Use With (Application) Laboratory evaporation, pharmaceutical, chemical, environmental, evaporation feed, food, beverage, academia, education analysis
    Temperature Range (English) 68° to 203°F
    Volume (Metric) 50 to 4000mL
    Depth (Metric) 42cm
    Capacity (Metric) 4000mL
    Includes 1L Evaporating and receiving flask, vertical condenser
    Electrical Requirements 100/120V 50/60Hz
    Hertz 50/60Hz
    Voltage 100/120V
    Width (Metric) 60cm
    Model RII
    Speed 20 to 280rpm
    Temperature Range (Metric) 20° to 95°C
    Product Type Glass Assembly C Rotary Evaporator
    Wattage 1700w
    Depth (English) 16.53 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit