Learn More
BUCHI Vacuum Seal WD-26 for Rotavapors™ R-210/215 and RII Available on GSA/VA Contract for Federal Government customers only.
Convenient and efficient rotary evaporation. Vacuum Seal WD-26 for Rotavapors™ R-210/215 and RII enables the user to realize the highest expectations in convenience and versatility.
Manufacturer:  BUCHI 048021
Catalog No. 05-422-873
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Rotavapor advantages:
  • Convenient - Comfortable flask and vapor duct removal thanks to easy to use Combi-Clip User-friendly operation due to ergonomic electric or manual lift Immediate adjustment of heating temperature and rotation speed with intuitively positioned control knobs
  • Extendable and versatile - Easy plug & play extension to a fully integrated system including a central interface, vacuum pump and recirculating chiller according to your needs Choice between 5 L high-temperature and 1 L fast-heating bath Wide range of beneficial accessories available: seven different glass assemblies, safety shield, lid for heating bath, automatic distillation and automatic foam detection
  • Reproducible - Precise operation control with integrated digital display of heating temperature, rotation speed and lift position Traceability of all process parameters including data recording and charting with the optional Interface I-300 Pro Instant alert with push notification on mobile devices in case of process deviation (with Interface I-300 / I-300 Pro only)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Low abrasion vacuum seal
Rotavapors™ R-210/215 and RII
Vacuum Seal WD-26
PTFE