Buckeye Fire Equipment ABC Fire Extinguishers

Multipurpose extinguishing agent that is suitable for use on Class A, Class B and Class C fires

Manufacturer: BUCKEYE FIRE EQUIPMENT CO 12120

 View more versions of this product

Catalog No. 19-120-828

  Check Availability
Add to Cart

Disclaimers:

Not recommended for use on sensitive electronic equipment or aircraft structure.


    Specifications

    Capacity 20 lb., 9.07kg
    Discharge Time 26 sec.
    Height (English) 21.25 in.
    Height (Metric) 53.9cm
    Includes Wall bracket
    Length (English) 7.5 in.
    Length (Metric) 19.05cm
    Material Steel
    Model 20S ABC
    Range 21 ft.
    Reagent Type ABC dry chemical
    UL Rating 6A:120B:C
    Capacity (English) 20 lb.
    Capacity (Metric) 9.07kg
    Product Type Fire extinguisher

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit