Learn More
BUD13 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 201631AP
Catalog No. 50-173-2739
Add to cart
Description
Description
BUD13 is a protein coding gene involved in pre-mRNA splicing as component of the activated spliceosome.
Specifications
Specifications
BUD13
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BRD0
BUD13, BUD13 homolog, BUD13 homolog (S. cerevisiae), fSAP71
Rabbit
IgG
150 μL
-20°C
Primary
84811
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
BUD13
Liquid
BUD13
BUD13 Fusion Protein Ag13665
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human