Learn More
Budesonide, Micronized, EP, 97.5-102%, Spectrum™
Click to view available options
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
B1595, 51333-22-3, C25H34O6
Spectrum™ Budesonide, Micronized, EP is a glucocorticoid steroid anti-inflammatory used to treat certain bowel conditions. Spectrum Chemical EP products, similar to USP products, are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle