Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Budesonide, Micronized, EP, 97.5-102%, Spectrum™

Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
51333-22-3
C25H34O6
430.54
VOVIALXJUBGFJZ-VXKMTNQYSA-N
(1S,2S,4R,6R,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
CCC[C@@H]1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
51333-22-3
C25H34O6
430.54
VOVIALXJUBGFJZ-VXKMTNQYSA-N
(1S,2S,4R,6R,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
CCC[C@@H]1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

B1595, 51333-22-3, C25H34O6

Spectrum™ Budesonide, Micronized, EP is a glucocorticoid steroid anti-inflammatory used to treat certain bowel conditions. Spectrum Chemical EP products, similar to USP products, are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Loss on Drying 0.5%
Quantity 5 g
Packaging Amber Glass Bottle
CAS Max % 100%
Percent Purity 97.5-102%