Learn More
New England Biolabs, Inc. NEBuffer™ 2.1 – 5 ml
SDP
Supplier:  New England Biolabs, Inc. B7202S
Encompass
New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers. Occasionally, an enzyme has specific buffer requirements not met by one of the four standard NEBuffers, in which case the enzyme is supplied with its own unique NEBuffer.
Catalog No. NC0458501
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.