Buffer HF-Improved, Alfa Aesar™

Used For Pretreatment of Planar Silicon Devices When Plating With Nickel Plating Solution Products 44069 and 44070

Manufacturer: Alfa Aesar 44627K2

Quantity 1L

 View more versions of this product

Catalog No. AA44627K2

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid
    MDL Number MFCD00132734
    Chemical Name or Material Buffer HF-Improved

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 1L
    DOT Information DOT Class: 8, Packaging Group: III
    Density 1.12g/cm3 at 20°C
    MDL Number MFCD00132734
    Health Hazard 1 H301-H311-H331-H314-H318: Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Buffer HF-Improved

    H301-H311-H331-H314-H318: Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit