Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer pH 7, Green, Reagents

Catalog No. p-200022077
Click to view available options
Quantity:
500 mL
4 L
10 L
20 L

Buffer pH 7, Green, Reagent, use for pH Meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Buffer pH 7, Green
CAS 7732-18-5, 7558-79-4, 7558-79-4
Boiling Point 100°C
Color Green
For Use With (Application) Calibration
For Use With (Equipment) pH Meter
pH 7
Physical Form Liquid
Quantity 10 L
Solution Type Buffer
Packaging Cubitainer
Specific Gravity 1.006
Sterility Non-sterile
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.