Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer, Precision Reference Standard, pH 4.000 ± 0.002 at 25°C (Color Coded Red), Ricca Chemical
Supplier: Ricca Chemical Company 150216
Specifications
Buffer, Precision Reference Standard | |
0°C | |
100°C | |
4 | |
500 mL | |
Miscible |
pH 4.00 ± 0.002 (Color Coded Red) | |
7647-01-0, 7732-18-5 | |
Red | |
Liquid | |
Clear PET-G Bottle |
Safety and Handling
ShelfLife : 24 months
DOTInformation : Not Regulated by DOT
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction