Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C (Color Coded Blue), Ricca Chemical

Supplier: Ricca Chemical Company 16011CT
Specifications
Buffer, Reference Standard | |
497-19-8, 144-55-8, 7732-18-5 | |
100.0, 0.53, 0.41 | |
Blue | |
0°C | |
4 L | |
Cubitainer |
pH 10.00 ± 0.01 at 25 deg C (Color Coded Blue) | |
97.0, 0.51, 0.39 | |
100°C | |
Liquid | |
10 | |
Miscible |
Safety and Handling
DOTInformation : Not Regulated by DOT
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title