Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C (Color Coded Blue), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
10 L
20 L
200 x 20 mL
Packaging:
Cubitainer
Natural Poly Bottle
Pack
Specifications
Specifications
| Chemical Name or Material | Buffer, Reference Standard |
| Name Note | pH 10.00 ± 0.01 at 25 deg C (Color Coded Blue) |
| Melting Point | 0°C |
| CAS | 497-19-8, 144-55-8, 7732-18-5 |
| Boiling Point | 100°C |
| CAS Min % | 97.0, 0.51, 0.39 |
| Color | Blue |
| CAS Max % | 100.0, 0.53, 0.41 |
| Concentration | 1X |
| For Use With (Application) | Calibration |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction