Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer, Reference Standard, pH 10.00 (Color Coded Blue), Ricca Chemical
$32.75 - $694.00
Specifications
Chemical Name or Material | Buffer, Reference Standard |
---|---|
Name Note | pH 10.00 ± 0.01 at 25 deg C (Color Coded Blue) |
CAS | 497-19-8, 144-55-8, 7732-18-5 |
CAS Min % | 97.0, 0.51, 0.39 |
CAS Max % | 100.0, 0.53, 0.41 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
1601-1
|
Ricca Chemical Company
16011 |
4L | Poly Bottle |
Each for $110.00
|
|
1601-16
![]() |
Ricca Chemical Company
160116 |
500mL | Poly Bottle |
Each for $32.75
|
![]() |
160125
|
Ricca Chemical Company
16012.5 |
10L | Cubitainer |
Each for $164.00
|
|
160120
|
Ricca Chemical Company
160120 |
20mL | Pouch (20/Box) |
Pack of 20 for $84.00
|
|
1601-1CT
|
Ricca Chemical Company
16011CT |
4L | Cubitainer |
Each for $112.00
|
|
1601-32
![]() |
Ricca Chemical Company
160132 |
1L | Poly Bottle |
Each for $55.50
|
![]() |
1601-5
![]() |
Ricca Chemical Company
16015 |
20L | Cubitainer |
Each for $223.00
|
![]() |
16-012-00
|
Ricca Chemical Company
160120B |
200 x 20mL | Individual use pouches |
Each for $694.00
|
|
Specifications
Buffer, Reference Standard | |
497-19-8, 144-55-8, 7732-18-5 | |
100.0, 0.53, 0.41 |
pH 10.00 ± 0.01 at 25 deg C (Color Coded Blue) | |
97.0, 0.51, 0.39 |
Safety and Handling
DOTInformation : Not Regulated by DOT
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title