Buffer, Reference Standard, pH 2.00 0.01 at 25°C, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 14932.5

Packaging Cubitainer™
Quantity 10L

 View more versions of this product

Catalog No. 1493-25

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7447-40-7 (Potassium Chloride),7647-01-0 (Hydrochloric Acid),7732-18-5 (Water, Deionized)
    Physical Form Liquid
    Chemical Name or Material Buffer, Reference Standard, pH 2.00± 0.01 at 25°C

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 10L
    Boiling Point 100°C
    DOT Information Not Regulated by DOT
    Shelf Life 24 months
    Melting Point 0°C
    pH 2
    Solubility Miscible
    CAS 7447-40-7 (Potassium Chloride),7732-18-5 (Water, Deionized),7647-01-0 (Hydrochloric Acid)
    Packaging Cubitainer™
    Chemical Name or Material Buffer, Reference Standard, pH 2.00± 0.01 at 25°C

    24 months

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit