Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, Reference Standard, pH 3.56 0.01 at 25°C, Ricca Chemical

Manufacturer:  Ricca Chemical Company 149816

 View more versions of this product

Catalog No. 1498-16


Add to cart

Description

Description

Specifications

Specifications

Buffer, Reference Standard
877-24-7, 7647-01-0, 7732-18-5
100.0, 1.04, 0.06
Liquid
Poly Natural Bottle
3.56
500 mL
pH 3.56± 0.01 at 25°C
97.0, 1.0, 0.06
Colorless Liquid
Reference Standard
3.55 to 3.57
Colorless
Non-sterile
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit