Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer, Reference Standard, pH 7.00 ± 0.01 at 25°C (Color Coded Green), Ricca Chemical
Supplier: Ricca Chemical Company 1551.41CT
Specifications
Buffer, Reference Standard | |
97.0, 0.87, 0.48 | |
100.0, 0.9, 0.5 | |
4 L |
7558-79-4, 7778-77-0, 7732-18-5 | |
Green | |
Liquid | |
Cubitainer |
Safety and Handling
ShelfLife : 24 months
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction