Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer, Reference Standard, pH 7.00 ± 0.01 at 25 deg C (Color Coded Yellow), Ricca Chemical
$31.50 - $695.00
Specifications
Chemical Name or Material | Buffer, Reference Standard |
---|---|
Name Note | pH 7.00 ± 0.01 at 25 deg C (Color Coded Yellow) |
CAS | 7558-79-4, 7778-77-0, 7732-18-5 |
CAS Min % | 97.0, 0.87, 0.48 |
CAS Max % | 100.0, 0.91, 0.5 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
1551-1
![]() |
Ricca Chemical Company
15511 |
4L | Plastic |
Each for $111.00
|
![]() |
1551-16
![]() |
Ricca Chemical Company
155116 |
500mL | Plastic |
Each for $31.50
|
![]() |
155120
|
Ricca Chemical Company
155120 |
20mL | Pouch (20/Box) |
Pack of 20 for $85.00
|
|
1551-32
|
Ricca Chemical Company
155132 |
1L | Plastic |
Each for $53.50
|
|
15511CT
![]() |
Ricca Chemical Company
15511CT |
4L | Cubitainer™ |
Each for $111.00
|
![]() |
1551-5
|
Ricca Chemical Company
15515 |
20L | Cubitainer™ |
Each for $216.00
|
|
1551-25
|
Ricca Chemical Company
15512.5 |
10L | Cubitainer™ |
Each for $160.00
|
|
15-512-00
|
Ricca Chemical Company
155120B |
200 x 20mL | Individual use pouches |
Each for $695.00
|
|
Specifications
Buffer, Reference Standard | |
7558-79-4, 7778-77-0, 7732-18-5 | |
100.0, 0.91, 0.5 |
pH 7.00 ± 0.01 at 25 deg C (Color Coded Yellow) | |
97.0, 0.87, 0.48 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title