Learn More
Supply SolutionsSupplier Diversity Partner Hach - Buffer Solution, Ph 7.00, 500Ml (Color Coded Yellow) (Nist) DFS Item
Manufacturer:  Supply SolutionsSupplier Diversity Partner HACH 2283549
Buffer solutions are ideal for laboratories doing high volume pH testing. These are color coded for easy identification. pH is ± 0.02 at 25°C. The 20 L package comes with a collapsible inner bladder in a White corrugated Cardboard box. These are NIST certified.; Parameter: ; Method Name: ; Number of tests: ; Description: Buffer Solution, (color-coded Yellow) (NIST); Concentration: 7.00 aprox. 0.02 pH; Volume: 500 mL; Platform : Solution
Catalog No. 50-476-308