Learn More
Supply SolutionsSupplier Diversity Partner Hach - Buffer Solution Red pH4.01 500ml; Parameter: pH; Method Name: ; Number of tests: ; Description: Buffer Solution, (color-coded Red) (NIST); Concentration: 4.01 aprox. 0.02 pH; Volume: 500 mL DFS Item
Manufacturer:  Supply SolutionsSupplier Diversity Partner HACH 2283449
Buffer solutions are ideal for laboratories doing high volume pH testing. These are color coded for easy identification. pH is ± 0.02 at 25°C. The 20 L package comes with a collapsible inner bladder in a White corrugated Cardboard box. These are NIST certified.; Parameter: pH; Method Name: ; Number of tests: ; Description: Buffer Solution, (color-coded Red) (NIST); Concentration: 4.01 aprox. 0.02 pH; Volume: 500 mL; Platform : Solution
Catalog No. 50-476-306