Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Buffer solution pH 10.0 (20°C), Borax / sodium hydroxide, Honeywell Fluka™

Click to view available options
Quantity:
100mL
500mL
1L
6 x 1L

Borax / sodium hydroxide

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Name or Material Buffer solution pH 10.0 (20°C)
Name Note Borax / sodium hydroxide
CAS 1310-73-2,1330-43-4,7732-18-5
CAS Max % <0.1000%,≤0.5000%
CAS Min % ≥0.2500%
Physical Form Buffer Solution
Packaging Poly Bottle
Boiling Point 100°C
Quantity 6 x 1L