Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Buffer solution pH 7.0 (20°C), Green colored, potassium dihydrogen phosphate / disodium hydrogen phosphate, with fungicide, Honeywell Fluka™

Click to view available options
Quantity:
500mL
1L
5L
6 x 500mL
10L
6 x 1L
1336-21-6
H5NO
35.046
MFCD00146206
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]
1336-21-6
H5NO
35.046
MFCD00146206
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]

Green colored, potassium dihydrogen phosphate / disodium hydrogen phosphate, with fungicide

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
UN Number NONH for all modes of transport
pH 7.0 (20°C)
Quantity 500mL
Chemical Name or Material Buffer solution