Alfa Aesar™ Buffer solution, pH 4.0, Coloured Red, Alfa Aesar™, Specpure™

Manufacturer: Alfa Aesar™ 38713K2

Quantity 1L

 View more versions of this product

Catalog No. AA38713K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1310-73-2,7778-77-0,7732-18-5
    MDL Number MFCD00146252
    Chemical Name or Material Buffer solution

    Specifications

    Color Red
    Physical Form Liquid
    Quantity 1L
    Appearance Red liquid
    MDL Number MFCD00146252
    CAS 1310-73-2,7778-77-0,7732-18-5
    Concentration or Composition (by Analyte or Components) Sodium hydroxide: 0.7%; Potassium dihydrogen phosphate: 0.7%; water: 98.6%
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Chemical Name or Material Buffer solution

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit