Learn More
Buffer solution pH 10.00 (+/-0.024 @ 25oC) Colored Blue Specpure NIST Traceable, Thermo Scientific Chemicals
$56.00 - $91.30
Specifications
Chemical Name or Material | Buffer solution |
---|---|
CAS | 1310-73-2, 7778-77-0, 7732-18-5 |
Color | Yellow |
Physical Form | Liquid |
MDL Number | MFCD00146206 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA38709K2
|
Thermo Scientific Chemicals
038709K2 |
1 L |
Each for $56.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AA38709K7
|
Thermo Scientific Chemicals
038709K7 |
4 L |
Each for $91.30
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Buffer solution (pH 10.00) is used as a means of keeping pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution | |
Yellow | |
MFCD00146206 | |
Miscible with water. |
1310-73-2, 7778-77-0, 7732-18-5 | |
Liquid | |
23 hPa (17mm Hg) at 20°C | |
Sodium hydroxide: 0.7%; Potassium dihydrogen phosphate: 0.7%; water: 98.6% |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.