Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution Duracal pH 9.21 (25°C),NIST/PTB reference materials, DKD certified, Honeywell Fluka™

250 mL, certified buffer solution traceable to NIST/PTB reference materials, DKD certified

$56.70

Specifications

Chemical Name or Material Buffer Solution Duracal Ph 9.21 (25°C)
Color Blue
Name Note Certified buffer solution traceable to NIST/PTB reference materials, DKD certified
UN Number NONH for all modes of transport
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
60-022-06
View Documents
Honeywell-Fluka
242147250ML
250mL
Each for $56.70
Add to cart
 
60-022-05
View Documents
Honeywell-Fluka
242146500ML
500mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Buffer Solution Duracal Ph 9.21 (25°C)
Blue
Certified buffer solution traceable to NIST/PTB reference materials, DKD certified
NONH for all modes of transport
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit