Learn More
Buffer solution pH 10.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals
$36.25 - $86.00
Specifications
Chemical Name or Material | Buffer solution |
---|---|
Name Note | pH 10.00 (±0.01 at 25°C), No Color, NIST Traceable |
CAS | 1336-21-6 |
Molecular Formula | H5NO |
MDL Number | MFCD00066650 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA38710AP
|
Thermo Scientific Chemicals
038710AP |
500 mL |
Each for $36.25
Order in Bulk
Request bulk or custom formats |
|
|||||
AA38710K2
|
Thermo Scientific Chemicals
038710K2 |
1 L |
Each
for $45.65
Save 12% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA38710K7
|
Thermo Scientific Chemicals
038710K7 |
4 L |
Each for $86.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution | |
1336-21-6 | |
MFCD00066650 | |
N.O | |
14923 | |
Specpure™ |
pH 10.00 (±0.01 at 25°C), No Color, NIST Traceable | |
H5NO | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
35.05 | |
CHEBI:18219 | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Warning
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.