Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 10.00, Color-Coded Blue, Mettler Toledo™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
500mL
4L
20L

Chemical Identifiers

Buffer Solution, pH 10.00
  • Quality solutions for accurate results
  • Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions
  • Each standard solution comes with a certificate of analysis, providing traceability to NIST standards

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

CAS 7732-18-5
Physical Form Liquid
Quantity 20L