Learn More
Buffer solution, pH 11.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 042424K7
Description
Buffer solution (pH 11) is used as a means of keeping pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution | |
Colorless | |
4 L | |
Odorless | |
Miscible with water. | |
N.O | |
35.05 | |
CHEBI:18219 |
1336-21-6 | |
Liquid | |
H5NO | |
MFCD00066650 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
azanium;hydroxide | |
14923 |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only