Buffer solution, pH 2.00 (±0.01 at 25deg.C), No Color, NIST Traceable, Alfa Aesar™, Specpure™

For Calibration of pH Meters

Manufacturer: Alfa Aesar 40447K7

Quantity 4L

 View more versions of this product

Catalog No. AA40447K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7447-40-7,7647-01-0,7732-18-5
    Physical Form Liquid
    MDL Number MFCD00146252
    Chemical Name or Material Buffer solution

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 4L
    CAS 7447-40-7,7647-01-0,7732-18-5
    Concentration or Composition (by Analyte or Components) Potassium chloride: 0.7%; Hydrochloric acid: 0.7%; water: 98.6%
    Appearance Colorless liquid
    Health Hazard 1 H318-H315: Causes serious eye damage. Causes skin irritation.
    Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste.
    Traceability to NIST Yes
    MDL Number MFCD00146252
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Chemical Name or Material Buffer solution

    In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste.

    H318-H315: Causes serious eye damage. Causes skin irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit