Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution pH 4.0 at 25°C, Color Coded Red, Certified, LabChem™

Catalog No. LC122801 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Cubitainer™
Pack
Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC122801 500 mL Poly Bottle
LC1228020 20 x 20 mL Pack
LC122802 1 L Poly Bottle
LC122804C 4 L Cubitainer™
LC122804 4 L Poly Bottle
LC122806 10 L Cubitainer™
LC122805 20 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. LC122801 Supplier LabChem, Inc. Supplier No. LC122801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Buffer Solution pH 4.0 at 25°C
Name Note Color Coded Red
Color Red
CAS Max % 98.935, 1.02, 0.04, 0.005
CAS 877-24-7, 50-00-0, 67-56-1
Decomposition Information Formaldehyde; Carbon monoxide; Carbon dioxide
Density 1g/mL
pH 4.00 to 4.01
Physical Form Liquid
Quantity 500 mL
Identification Passes Test
Linear Formula KHC8H4O4
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 204.23
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.