Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 4.00, Color-Coded Pink, Mettler Toledo™

Click to view available options
Quantity:
500mL
4L
877-24-7
C8H5KO4
204.222
IWZKICVEHNUQTL-UHFFFAOYSA-M
Potassium Biphthalate
23676735
potassium;2-carboxybenzoate
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
877-24-7
C8H5KO4
204.222
IWZKICVEHNUQTL-UHFFFAOYSA-M
Potassium Biphthalate
23676735
potassium;2-carboxybenzoate
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
  • Quality solutions for accurate results
  • Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions
  • Each standard solution comes with a certificate of analysis, providing traceability to NIST standards
Formula Weight 204.23
Physical Form Liquid
Quantity 4L
Chemical Name or Material Buffer Solution, pH 4.00