Alfa Aesar™ Buffer solution, pH 5.00 (+/-0.01 at 25°C), No Color, Specpure™, NIST Traceable

Manufacturer: Alfa Aesar™ 42417AP

Quantity 500mL

 View more versions of this product

Catalog No. AA42417AP

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00147253
    Chemical Name or Material Buffer solution, pH 5.00 (±0.01 at 25°C), NIST Traceable
    Grade Specpure™

    Specifications

    Quantity 500mL
    Physical Form Liquid
    MDL Number MFCD00147253
    Color No Color
    TSCA TSCA
    Chemical Name or Material Buffer solution, pH 5.00 (±0.01 at 25°C), NIST Traceable
    Grade Specpure™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit