Learn More
Buffer Solution, pH 6.0, Acetate, For Aluminum, APHA, Spectrum™
Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner B187500ML
Catalog No. 18-609-332
Add to cart
Specifications
Specifications
Chemical Solution
Poly Bottle
500 mL
6131-90-4, 64-19-7, 7732-18-5
6.00 ± 0.02
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.