Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer Solution pH 7.0 at 25°C, Color Coded Yellow, Certified, LabChem™
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Cubitainer™
Pack
Poly Bottle
Specifications
Specifications
| Chemical Name or Material | Buffer Solution pH 7.0 at 25°C |
| Name Note | Color Coded Yellow |
| Color | Yellow |
| CAS Max % | 99.24, 0.68, 0.08 |
| CAS | 7778-77-0, 1310-73-2 |
| Decomposition Information | Phosphorus oxides; Sodium oxide |
| pH | 7.00 ±0.01 |
| Physical Form | Liquid |
| Quantity | 4 L |
| Identification | Passes Test |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction