Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution pH 7.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable

Catalog No. AA38712K7
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38712K7 4 L
AA38712AP 500 mL
AA38712K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA38712K7 Supplier Thermo Scientific Chemicals Supplier No. 038712K7
Add to Cart

CAS: 1336-21-6 | H5NO | 35.05 g/mol

Used to determine pH of solutions in various industrial applications. Buffer solutions of known and accurate pH value allows to adjust testing system to measure the samples with unknown pH more precisely and accurately.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution
CAS 1336-21-6
Color Colorless
Physical Form Liquid
Quantity 4 L
Molecular Formula H5NO
Odor Odorless
MDL Number MFCD00066650
Traceability to NIST Yes
Solubility Information pH 7.00 buffers are in solution form. Miscible with water.
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name ammonium hydroxide
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Grade Specpure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.