Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 7.00, Color-Coded Green, Mettler Toledo™

Click to view available options
Quantity:
500mL
4L
7778-77-0
H2KO4P
136.084
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
CHEBI:63036
potassium;dihydrogen phosphate
OP(=O)(O)[O-].[K+]
7778-77-0
H2KO4P
136.084
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
CHEBI:63036
potassium;dihydrogen phosphate
OP(=O)(O)[O-].[K+]
  • Quality solutions for accurate results
  • Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions
  • Each standard solution comes with a certificate of analysis, providing traceability to NIST standards
Physical Form Liquid
NIST NIST Traceable
Quantity 4L
Chemical Name or Material Buffer Solution, pH 7.00