Learn More
Buffer Solution, pH 7.38, Reference Standard, Spectrum™
Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner B198500ML
 View more versions of this product
Catalog No. 18-609-346
Add to cart
Specifications
Specifications
Chemical Solution
Poly Bottle
500 mL
7778-77-0, 1310-73-2, 7732-18-5
7.38 ± 0.02
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.