Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 7.40, Reference Standard, Spectrum™

$22.05 - $76.65

Specifications

pH 7.40 ± 0.01
CAS 7778-77-0, 1310-73-2, 99-76-3
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18-609-349
View Documents
Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
B301500MLPL
500 mL Poly Bottle Each for $22.05
Add to cart
 
18-609-350
View Documents
Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
B3011LT
1 L Poly Bottle Each for $38.85
Add to cart
 
18-609-351
View Documents
Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
B3014LT
4 L Poly Bottle Each for $76.65
Add to cart
 
Specifications

Specifications

7.40 ± 0.01
7778-77-0, 1310-73-2, 99-76-3

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit