Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 7, Alfa Aesar™, Specpure™

Click to view available options
Quantity:
500mL
1L
4L
1336-21-6
H5NO
35.046
MFCD00146206
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]
1336-21-6
H5NO
35.046
MFCD00146206
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]
Appearance Yellow liquid
Solubility Information Miscible with water.
Physical Form Liquid
Grade Specpure™
Traceability to NIST Yes
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Color Green
Quantity 4L
Chemical Name or Material Buffer solution, NIST Traceable

TSCA : No

Recommended Storage : Ambient temperatures