Learn More
Buffer solution pH 7.00 (+/-0.024 @ 25oC) Colored Green Specpure NIST Traceable, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 038711AP
Description
Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution | |
1336-21-6 | |
Liquid | |
H5NO | |
Yes | |
Miscible with water. | |
N.O | |
35.05 | |
CHEBI:18219 |
NIST Traceable | |
Green | |
500 mL | |
MFCD00066650 | |
23 hPa (17mm Hg) at 20°C | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
azanium;hydroxide | |
14923 | |
Specpure™ |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only