Buffer solution, pH 8 (±0.01 at 25deg.C), No Color, NIST Traceable, Alfa Aesar™, Specpure™

Manufacturer: Alfa Aesar 42420K7

Quantity 4L

 View more versions of this product

Catalog No. AA42420K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1310-73-2,7778-77-0,7732-18-5
    Physical Form Liquid
    MDL Number MFCD00134130
    Chemical Name or Material Buffer solution

    Specifications

    Physical Form Liquid
    Quantity 4L
    Color Yellow
    MDL Number MFCD00134130
    CAS 1310-73-2,7778-77-0,7732-18-5
    Concentration or Composition (by Analyte or Components) Sodium hydroxide: 0.7%; Potassium dihydrogen phosphate: 0.7%; Water: 98.6%
    Appearance Yellow liquid
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Chemical Name or Material Buffer solution

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit