Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BugBuster™ Plus Lysonase™ Kit

Combination of two reagents allows maximum recovery of active soluble protein from both Gram-negative and Gram-positive bacteria

$970.00

Specifications

Content And Storage Multiple storage requirements
Quantity 1 kit
View More Specs

Products 2
Catalog Number Mfr. No. Kit Contents Price Quantity & Availability  
Catalog Number Mfr. No. Kit Contents Price Quantity & Availability  
71-370-4
View Documents
MilliporeSigma™
713704
BugBuster Protein Extraction Reagent (500mL), Lysonase Bioprocessing Reagent (1mL)
Each for $970.00
Add to Cart
 
71-370-3
View Documents
MilliporeSigma™
713703
BugBuster Protein Extraction Reagent (100mL), Lysonase Bioprocessing Reagent (200μL) Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area. N/A
Description

Description

  • The BugBuster and rLysozyme efficiently lyse the bacterial cells while the Benzonase degrades nucleic acids to facilitate downstream processing
  • Use 5mL BugBuster and 10μL Lysonase per 1g cell paste
  • The kits provide sufficient reagents for protein extraction from 20g and 100g cell paste
Specifications

Specifications

Multiple storage requirements
1 kit
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit