Learn More
Bachem 5-Azido-pentanoic acid, 1g, 79583-98-5, MFCD11850108 DFS Item
Manufacturer:  Bachem 4059927.0001
Building block for "click chemistry". The azido group reacts with acetylenes in the presence of Cu/CuSO₄ yielding triazoles. This highly selective reaction can be applied for the conjugation of peptides with various molecules and for the cyclization of peptides. The azido moiety may also be considered as a protected amino group which is deblocked under reductive conditions.
Catalog No. 50-194-7399