Learn More
Bachem 1-Amino-4-chloro-2-butyne · HCl, 5g, 77369-59-6, MFCD00236918 DFS Item
Manufacturer:  Bachem 4010466.0005
Building block for the synthesis of enzyme inhibitors such as inhibitors of amine oxidases. Educt for the synthesis of derivatives of the growth inhibitor 2,6-diamino-4-hexynoic acid.
Catalog No. 50-194-7404