Bulbtronics™ EIKO Fluorescent Bulb: Shape T5

Manufacturer: Bulbtronics, Inc. 017385

 View more versions of this product

Catalog No. 14-395-821

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Base Style G5/Mini Bi Pin
    Color Blacklight Blue
    Diameter (English) Outer 0.62 in.
    Diameter (Metric) Outer 1.58cm
    For Use With (Equipment) Stratoliner model 1800, gene splicing equipment
    Length (English) 9 in.
    Length (Metric) 22.8cm
    Lighting Life 7500 hr.
    Shape T5 Shape
    Wattage 6w
    Light Source Fluorescent
    Product Type Linear Bulb

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit