Learn More
Bulbtronics™ Bulbtronics™ General Electric™ Germicidal Bulb
254nm in a variety of styles and dimensions
Manufacturer:  Bulbtronics™ 032558
 View more versions of this product
Catalog No. 14-395-849
Add to cart
Description
Description
  • Style: linear double ended
  • Wavelength: 254nm
Specifications
Specifications
FA8/slimline, Single Pin
0.62 in.
Aqua fine 3082/3084
91.4cm
9000 hr.
39w
Linear Double Ended Bulb
Preheat
1.58cm
36 in.
Germicidal
T5 Shape
254nm