Learn More
Bulbtronics™ General Electric™ Halogen Lamp: MR16
Manufacturer:  Bulbtronics™ 001191/DEL
 View more versions of this product
Catalog No. 14-395-465
Add to cart
Specifications
Specifications
GX5.3/2 Pin
3150K
5.08cm
Surgical light, medical diagnostic
4.44cm
MR16 Shape
150w
Halogen Lamp
Base down to horizontal
2 in.
C6/Single Coil Transverse
1.75 in.
500 hr.
20V
Halogen